4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
Supplier: Matrix Scientific
011450-1G
102550-966EA
198.46
USD
102550-966
4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
Hazard: Irritant
Formula:
C₈H₁₀N₄ MW: 162.2 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00124667 CAS Number: 41601-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...