Piperidin-1-ylacetic acid hydrochloride
Supplier: Matrix Scientific
011387-1G
102832-842EA
36.92
USD
102832-842
Piperidin-1-ylacetic acid hydrochloride
Piperidin-1-ylacetic acid hydrochloride
Hazard: Irritant
Formula:
C₇H₁₄ClNO₂ MW: 179.65 g/mol Melting Pt: 207…209 °C |
MDL Number:
MFCD01701566 CAS Number: 3235-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...