2-Amino-5,6-dihydro-4H-benzothiazol-7-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5,6-dihydrobenzo[d]thiazol-7(4H)-one

010114-5G 010114-1G
102832-814EA 60 USD
102832-814 102832-816
2-Amino-5,6-dihydro-4H-benzothiazol-7-one ≥95%
2-Amino-5,6-dihydro-4H-benzothiazol-7-one

Hazard: Irritant

Formula: C₇H₈N₂OS
MW: 168.22 g/mol
Melting Pt: 269…270 °C
MDL Number: MFCD00460519
CAS Number: 17583-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR