5-(Hydroxymethyl)furan-2-carboxylic Acid
Supplier: Matrix Scientific
Synonyms:
Sumiki's acid
, 5-(Hydroxymethyl)-2-furoic acid
, 5-Hydroxymethyl-2-furancarboxylic acid
009897-1G
102550-948EA
36.92
USD
102550-948
5-(Hydroxymethyl)furan-2-carboxylic Acid
5-(Hydroxymethyl)furan-2-carboxylic Acid
Formula:
C₆H₆O₄ MW: 142.11 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD03274472 CAS Number: 6338-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...