5-Bromo-7-fluoroindole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-7-fluoro-1H-indole
009819-250MG
009819-1G
102663-578EA
461.54
USD
102663-578
102663-576
5-Bromo-7-fluoroindole ≥95%
5-Bromo-7-fluoroindole
Hazard: Irritant/Refrigerate
Formula:
C₈H₅BrFN MW: 214.04 g/mol |
MDL Number:
MFCD04037876 CAS Number: 883500-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...