3-(4-Methylpiperazin-1-yl)propylamine

Supplier: Matrix Scientific

Synonyms: 1-(3-Aminopropyl)-4-methylpiperazine

008505-1G
102550-924EA 184.62 USD
102550-924
3-(4-Methylpiperazin-1-yl)propylamine
3-(4-Methylpiperazin-1-yl)propylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₉N₃
MW: 157.26 g/mol
Boiling Pt: 113…114 °C (13 mmHg)
Density: 0.924
MDL Number: MFCD00014616
CAS Number: 4572-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR