4-Chloro-8-methoxy-2-methylquinoline

Supplier: Matrix Scientific

007931-1G
102550-910EA 118.45 USD
102550-910
4-Chloro-8-methoxy-2-methylquinoline
4-Chloro-8-methoxy-2-methylquinoline

Hazard: Irritant

Formula: C₁₁H₁₀ClNO
MW: 207.66 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00272401
CAS Number: 64951-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR