1-(5-Bromopyrimidin-2-yl)-[1,4]-diazepane
Supplier: Matrix Scientific
Synonyms:
1-(5-Bromopyrimidin-2-yl)homopiperazine
007333-1G
007333-5G
102550-898EA
153.85
USD
102550-898
102550-900
1-(5-Bromopyrimidin-2-yl)-[1,4]-diazepane
1-(5-Bromopyrimidin-2-yl)-[1,4]-diazepane
Hazard: Irritant
Formula:
C₉H₁₃BrN₄ MW: 257.14 g/mol Melting Pt: 203…207 °C |
MDL Number:
MFCD03426069 CAS Number: 849021-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...