Methyl-6-fluorosalicylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 2-fluoro-6-hydroxybenzoate

003820-1G
102550-830EA 90.77 USD
102550-830
Methyl-6-fluorosalicylate ≥98%
Methyl 6-fluorosalicylate

Hazard: Irritant

Formula: C₈H₇FO₃
MW: 170.1378 g/mol
Boiling Pt: 58…60 °C (0.3 mmHg)
MDL Number: MFCD00153207
CAS Number: 72373-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR