1,2,3,3,3-Pentafluoropropene ≥97%
Supplier: Matrix Scientific
001547-25G
001547-5G
102550-798EA
947.69
USD
102550-798
102550-796
1,2,3,3,3-Pentafluoropropene ≥97%
1,2,3,3,3-Pentafluoropropene
Hazard: Nonflammable Gas
Formula:
C₃HF₅ MW: 132.03 g/mol Boiling Pt: –20 °C |
MDL Number:
MFCD02262202 CAS Number: 2252-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...