Anti-Human IFN-gamma Antibody
08P990851
10497-066EA 309.02 USD
10497-066
Anti-Human IFN-gamma Antibody
Produced from sera of rabbits pre-immunized with highly pure recombinant Human IFN-γ. Anti-Human IFN-γ specific antibody was purified by affinity chromatography employing immobilized Human IFN-γ matrix.
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/mL.