Anti-IFN-γ Rabbit Antibody (Biotin)
08P990891
10497-068EA 309.02 USD
10497-068
Anti-IFN-γ Rabbit Antibody (Biotin)
Produced from sera of rabbits pre-immunized with highly pure recombinant Human IFN-γ. Anti-Human IFN-γ specific antibody was purified by affinity chromatography and then biotinylated.
Centrifuge vial prior to opening. Reconstitute in sterile PBS containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL.