Would you like to log in again?
Marvel Scientific SA24RAS ADA Height All-Refrigerator, Aga Marvel
Supplier: Marvel Scientific
SA24RAS4LB SA24RAS4RW SA24RAS5RS SA24RAS4RB SA24RAS4LW SA24RAS5LS
These solid door undercounter ADA height all-refrigerators are less than 31 1/8" high, allowing them to be built-in under ADA height 34" counter tops
These ADA height all-refrigerators feature the exterior door-front-mounted MicroSentry II Scientific TM temperature controller for accurate control and convenient monitoring of the storage temperatures within the cabinet interior, without opening the door. The MicroSentry II Scientific TM temperature controller allows the user to select a set point temperature between 1 and 7 degrees C and establish high and low temperature alarm values around the set point. The user can also set-up the length of the time period an alarm condition must be present before the audible and visual alarms will be activated, thereby reducing nuisance alarms. The MicroSentry II Scientific TM controller also has a door open alarm, to notify the refrigerator user of this most common cause for out of range temperatures before they occur. Other features of the MicroSentry II Scientific TM temperature controller include, highest and lowest temperature history record, built-in diagnostics, and control keypad lock-out to prevent unintended control inputs. The MicroSentry II Scientific TM temperature controller in these units can maintain the stored product temperature inside the unit interior within +2 and -2 degrees C of the set point temperature.
The interiors of these ADA height all-refrigerators feature an advanced fan forced air cooling system, a flat inner door panel without shelves and three full depth perforated metal shelves that are cantilever mounted on the rear wall of the interior and are adjustable in 1.5" increments. Additional standard features include a probe port on the rear wall of the interior, interior light, automatic defrost without an external heat cycle, a positive-close door hinge system, hospital grade plug, four adjustable leveling feet installed in the cabinet base, and a CSA Certification.
They feature an exterior door-front-mounted MicroSentry II Scientific TM temperature control with a digital temperature display and user interface. They incorporate an advanced forced air evaporator system for more uniform interior temperatures and more precise maintenance of their set point temperature. In addition, they feature a fan forced air cooled condenser and large capacity compressor, which allow these undercounter units to be built into casework with zero clearance required for cooling air at the back, top and sides, without compromising their cooling performance. They can also be installed freestanding or stacked in pairs.
Certifications: CSA Approved
Ordering information: Door lock included.
ORDER
Expand All / Collapse All
Volume Color Door Type Supplier No. VWR Catalog Number Unit Price Quantity
130 L (4.6 cu. ft.) Stainless Cabinet/Stainless Door Left Hinge SA24RAS5LS 75796-354 Each Retrieving
CSA Certified
130 L (4.6 cu. ft.) Stainless Cabinet/Stainless Door Right Hinge SA24RAS5RS 75796-352 Each Retrieving
CSA Certified
130 L (4.6 cu. ft.) White Cabinet/White Door Left Hinge SA24RAS4LW 75796-262 Each Retrieving
CSA Certified
130 L (4.6 cu. ft.) White Cabinet/White Door Right Hinge SA24RAS4RW 75796-260 Each Retrieving
CSA Certified
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Electrical 115V/60Hz
Refrigerant R134a
SERVICES
Expand All / Collapse All
VWR® Customer On-site Calibration, Validation and PM Services
Description Details Includes Service Coverage Out-of-Service Coverage Supplier No. VWR Catalog Number Unit Price Quantity
VWR® NIST On-Site Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. Calibration at a single point using NIST traceable standards. Upon completion, a calibration form will be provided with “as found” and “as left” measurements. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-880 Each Retrieving
VWR® NIST On-Site PM and Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. PM and Calibration service includes cleaning of seals, fans, and gaskets. The chamber is inspected interiorly and exteriorly. After PM, technicians will calibrate the chamber as described in Calibration section. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-888 Each Retrieving
VWR® NIST Traceable Validate (IOQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. Validation (IQ/OQ) includes a standardized protocol procedure, calibration, tuning, IQ (verify design spec, system SOP, environmental conditions etc), and OQ (Perform empty chamber temperature mapping, open door and power loss testing). (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-950 Each Retrieving
VWR® NIST Traceable Validate (IOPQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. IQ/OQ will be performed as described in the (IQ/OQ) section. The PQ consists of loaded chamber temperature mapping for an additional 24 hours. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-942 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty