Inert PEEK Tubing, Restek
25067 25068 25066 25065 25320
10059-350EA 55.15 USD
10059-350 10059-348 10059-414 10059-346 10059-352
Inert PEEK Tubing, Restek
  • Replaces stainless steel, titanium, PTFE, or Tefzel® tubing
  • Less oxygen permeable and more temperature resistant (to 100 °C) than PTFE or Tefzel® tubing
  • Use with PEEK finger-tight or flangeless fittings.Use tubing ≤ 0.007" to 7,000 psi; tubing ≥ 0.010" ID to 5,000 psi