Quadruple Baffled Erlenmeyer Flask, Chemglass
Supplier: Chemglass
CLS-2093-0500 CLS-2093-2000 CLS-2093-0125 CLS-2093-0250 CLS-2093-1000
89209-378PK 301.51 USD
89209-378 89209-383 89209-381 89209-376 89209-374
Quadruple Baffled Erlenmeyer Flask, Chemglass
Quadruple baffled Erlenmeyer flasks are sterile, non-pyrogenic and manufactured with polycarbonate resin.
  • Graduations are molded for accuracy
  • New EZ-Tear packaging allows for easy access
Delivery information: Flasks are supplied with green filter caps.