Would you like to log in again?
SevenCompact S210 pH Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
30130864 30130863 30130865
Starting a measurement or calibration requires only a single keypress. Menu settings are intuitive, consisting of full sentences in 12 languages.
  • Truly intuitive operation
  • High–resolution color display
  • Automatic buffer recognition
  • All–round productivity from the start
The 4.3 inch color display with its large digits and well arranged icons makes the relevant data identifiable at a glance.
The automatic buffer recognition system allow you to choose the order in which to use buffers when calibrating. This way you quickly get through your routine calibrations without unnecessary error messages.
Certifications: UL listed and CSA certified.
Ordering information: S210 pH meter kit comes with the pH/mV meter, a uPlace electrode stand, and an electrode suited to your unique application. Start measuring as soon as it arrives!
ORDER
Expand All / Collapse All
Description Includes Supplier No. VWR Catalog Number Unit Price Quantity
SevenCompact S210–Kit InLab Expert Pro ISM electrode 30130863 75801-398 Each Retrieving Restricted
UL Listed CSA Certified
SevenCompact S210–Bio InLab Routine Pro ISM Electrode 30130864 75801-400 Each Retrieving Restricted
UL Listed CSA Certified
SevenCompact S210–U InLab Versatile Pro electrode 30130865 75801-390 Each Retrieving Restricted
UL Listed CSA Certified
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Resolution 0.001 / 0.01 / 0.1 (pH);
0.1 / 1 (mV);
0.1 C (ATC/MTC)
Interfaces USB A, USB B, RS232
Languages 12 Languages available
Measurement Range –2.000 (pH) [min.] to 20.000 (pH) [max.];
–2000.0 (mV) [min.] to 2000.0 (mV) [max.];
–5.0 C (ATC), –30 (MTC) [min.] to 130 C (ATC/MTC) [max.]
Ingress Rating IP54
Limit of Error Low range:
0.002 (pH);
0.2 (mV);
0.1 C (ATC/MTC)
SERVICES
Expand All / Collapse All
METTLER TOLEDO® Services
Description Supplier No. VWR Catalog Number Unit Price Quantity
IPac Standard Qualification (IQ/OQ), SevenCompact (Installation Pac) B39990007 10199-716 Each Retrieving
EQPac (IQ/OQ/PQ) Comprehensive Qualification, SevenExcellence (Installation Pac) B39990008 10199-718 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty