Pump Plate Only
S71818-10
470123-252EA 203.99 USD
470123-252
Pump Plate Only
This simple hand pump is convenient for very rapid work for either exhaust or compression, depending upon the nozzle to which it is connected.
The pump is about 25cm high, is sturdily built, and works on the same principle as an ordinary bicycle or hand pump except for having nozzles for both pressure and vacuum. The up stroke of the pump is effective in producing vacuum, while the down stroke produces the pressure from a different nozzle.