    Sort by:
  • Search Within Results
You Searched for: "neutralizing-solution"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  SOLON DISTRIBUTION CENTER
Description:   A pH adjusted solution of 1.5M sodium chloride, and 1.0M tris buffer in specially prepared water (pH 7.0).