J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Bismuth
Supplier: Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®
BAKR6445-04 BAKR6445-04
JT6445-4CS 374.09 USD
CAJT6445-4 JT6445-4
J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Bismuth
Standards Cationic Standards Bismuth Standards
The standard is prepared by the careful dilution of the metal or its salt, usually 99.99% pure as verified by arc emission spectrometry.
  • Packaged at a concentration of 1000 µg/ml
  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.