J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Copper
Supplier: Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®
BAKR6451-04 BAKR6451-04
JT6451-4CS 1117.54 USD
JT6451-4 CAJT6451-4
J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Copper
Standards Cationic Standards Copper Standards
The standard is prepared by the careful dilution of the metal or its salt, usually 99.99% pure as verified by arc emission spectrometry
  • Packaged at a concentration of 1,000µg/mL
  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.