Zinc AAS Standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
BAKR6474-04 BAKR6474-04
JT6474-4CS 235 USD
JT6474-4 CAJT6474-4
Zinc AAS Standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
These standards are prepared by the careful dilution of selected metals or salts, usually 99.99% pure as verified by arc emission spectrometry
  • Standards packaged at a concentration of 1,000µg/mL
  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.