9-Fluorenylmethyl chloroformate (Fmoc chloride), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 9H-Fluoren-9-ylmethyl chloroformate(9H-Fluoren-9-yl)methyl carbonochloridateFmoc-Cl9-Fluorenylmethyl chloroformateChloroformic acid 9-fluorenylmethyl ester, FMOCCl

M-Clarity™ quality level = MQ200

8.18203.8500 8.18203.8500
EM8.18203.8500EA 23 USD
EM8.18203.8500 CA8.18203.8500
9-Fluorenylmethyl chloroformate (Fmoc chloride), Sigma-Aldrich®
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula: C₁₅H₁₁ClO₂
MW: 258.7 g/mol
MDL Number: MFCD00001138
CAS Number: 28920-43-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR