Methylstyrene (mixture of isomers), Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.22098.9045 8.22098.9045
EM8.22098.9045EA 2268 USD
CA8.22098.9045 EM8.22098.9045
Methylstyrene (mixture of isomers), Sigma-Aldrich®
Methylstyrene (mixture of isomers)
Formula: C₉H₁₀
MDL Number: MFCD00168066
CAS Number: 25013-15-4
UN: 2618
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR