Triethylammonium chloride, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Triethylamine hydrochloride

M-Clarity™ quality level = MQ200

8.21135.9025 8.21135.9025
EM8.21135.9025EA 1396 USD
EM8.21135.9025 CA8.21135.9025
Triethylammonium chloride, Sigma-Aldrich®
Triethylammonium chloride
Formula: C₆H₁₆ClN
MW: 137.65 g/mol
Density: 0.7637 g/cm³ (20 °C)
MDL Number: MFCD00012500
CAS Number: 554-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR