Isophorone, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 3,5,5-Trimethylcyclohex-2-enone3,5,5-Trimethyl-2-cyclohexen-1-one

M-Clarity™ quality level = MQ200

8.04807.9026 8.04807.9026
EM8.04807.9026EA 900 USD
EM8.04807.9026 CA8.04807.9026
Isophorone, Sigma-Aldrich®
Isophorone
Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.922 g/cm³ (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00001584
CAS Number: 78-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR