2-Aminophenol, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Hydroxyaniline

M-Clarity™ quality level = MQ200

8.00419.9050 8.00419.9050
EM8.00419.9050EA 5389 USD
EM8.00419.9050 CA8.00419.9050
2-Aminophenol, Sigma-Aldrich®
2-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 235 °C (1013 hPa)
Density: 1.328 g/cm³ (20 °C)
Flash Pt: >175 °C
MDL Number: MFCD00007690
CAS Number: 95-55-6
UN: 2512
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR