3-Chlorophenol, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20321.0010 8.20321.0010
EM8.20321.0010EA 33 USD
EM8.20321.0010 CA8.20321.0010
3-Chlorophenol, Sigma-Aldrich®
3-Chlorophenol
Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: ∼214 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.245 g/cm³ (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00002256
CAS Number: 108-43-0
UN: 2020
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR