Dihexylamine, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20465.0050 8.20465.0050
EM8.20465.0050EA 65 USD
CA8.20465.0050 EM8.20465.0050
Dihexylamine, Sigma-Aldrich®
Dihexylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.788 g/cm³ (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00009521
CAS Number: 143-16-8
UN: 2927
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR