1-Dodecanethiol, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
n-Dodecyl mercaptan
, 1-Mercaptododecane
, n-Dodecanethiol
M-Clarity™ quality level = MQ200
8.20544.9011
8.20544.9011
EM8.20544.9011CS
12601
USD
CA8.20544.9011
EM8.20544.9011
1-Dodecanethiol, Sigma-Aldrich®
1-Dodecanethiol
Formula:
C₁₂H₂₆S MW: 202.4 g/mol Boiling Pt: 269…273 °C (1013 hPa) Melting Pt: –7 °C Density: 0.855 g/cm³ (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00004885 CAS Number: 112-55-0 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...