1-Hexanol, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Hexyl alcohol1-Hydroxy hexane1-Hexyl alcoholPentyl carbinol

M-Clarity™ quality level = MQ200

8.04393.9020 8.04393.9020
EM8.04393.9020EA 743 USD
EM8.04393.9020 CA8.04393.9020
1-Hexanol, Sigma-Aldrich®
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.8322 g/cm³ (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00002982
CAS Number: 111-27-3
UN: 2282
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/12/48/731248.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR