Butyric acid, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: n-Butyric acidButanoic acid

M-Clarity™ quality level = MQ200

8.00457.9050 8.00457.9050
EM8.00457.9050EA 1348 USD
EM8.00457.9050 CA8.00457.9050
Butyric acid, Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 163.5 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00002814
CAS Number: 107-92-6
UN: 2820
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR