SP Wilmad-LabGlass Reaction Flask Heating Mantles, Side Zip, SP Industries
Supplier: SP Industries
LG-8877-108 LG-8877-106 LG-8877-102 LG-8877-110 LG-8877-100 LG-8877-104
14200-718EA 592.17 USD
14200-718 14201-040 14200-640 14201-038 14200-526 14201-098
SP Wilmad-LabGlass Reaction Flask Heating Mantles, Side Zip, SP Industries
These all-glass constructed heating mantles are for use with LG-8010 and LG-8011 reaction flasks. They provide even heat distribution and prevent scorching of clear materials.
  • Mantles feature a 1.25" opening to accommodate bottom take-off tube and a zipper for easy on and off
  • Depth given is the interior measurement
  • All wattage is for 115 V circuits
Heating circuits have two short lead-in wires connected by a Hubbell plug to a 2-wire type SJT cord, 4 feet long.