Would you like to log in again?
IgG Rheumatoid Factor Mouse ELISA, BioVendor
Supplier: BioVendor
RSHAKRRG101R
Rheumatoid factors are autoantibodies against Fc region of of IgG and is found in 70–80% of patients suffering from chronic rheumatoid arthritis, and are considered to be closely related to its pathological syndrome.
  • Calibration Range: 15.6–1000 mU/mL
  • Limit of Detection: 0.78 mU/mL
  • Plasma-Citrate, Plasma-EDTA, Serum: 1 uL/well
Rheumatoid factors are autoantibodies against Fc region of of IgG and is found in 70–80% of patients suffering from chronic rheumatoid arthritis, and are considered to be closely related to its pathological syndrome. Experimental animal models with spontaneous autoimmune diseases similar to those in humans, and animals with artificially-induced inflammation have been used to elucidate the mechanism of autoimmune diseases and to search for potential new remedies. A representative model animal of spontaneous autoimmune diseases is MRL/lpr mouse. As MRL/lpr shows high incidence of lymph node tumor, nephritis, angitis, and arthritis, this animal strain is useful for studies of the mechanism of human autoimmune diseases including rheumatoid arthritis. Autoantibodies found in MRL/lpr serum are IgG type rheumatoid factor, IgM type rheumatoid factor, anti-ssDNA antibodies, anti-dsDNA antibodies, and anti-Sm antibody, etc. This kit enables quantification and comparison of IgG type rheumatoid factor with a calibration curve using standard antibody preparation.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
ORDER
Expand All / Collapse All
Description Size Regulatory Status Supplier No. VWR Catalog Number Unit Price Quantity
IgG Rheumatoid Factor Mouse ELISA 1 kit RUO RSHAKRRG101R 101424-360 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty