Would you like to log in again?
Draft Shield XP XS, Mettler Toledo
Supplier: Mettler Toledo
11131653
11278-494EA 816.1 USD InStock
11278-494
Draft Shield XP XS, Mettler Toledo
Shields Safety Shields
Protected weighing by minimizing air currents
  • Improve the quality of results by removing the influence of air currents
  • Faster settling times and more precise results
Add the Simple Draft Shield to a balance to create a chamber which protects both weighing pan and samples from spurious air currents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty