Would you like to log in again?
VWR® FILTRA-BAG® Blender Bags
Supplier: VWR International
EFT-1515A-VW1 EFT6090A-VW1
76196-374CS 1069.32 USD InStock
76196-374 89085-570
VWR® FILTRA-BAG® Blender Bags
Bags Blender Bags
FILTRA-BAG blender bags are designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid / solid substances.
  • Puncture-resistant
  • Durable fabrication
When lacing an aliquot onto growth media, it's very important that the volume of the aliquot be accurate and that it be free of solid particles. This prevents pipette blockage as well as uneven plate distribution. Polyethylene/nylon outer shell which practically eliminates the possibility of piercing the bag wall during blending.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty