Headspace Crimp Top Vials, SUN SRI™
Supplier: SUN SRi
405 034 200 898 500 550 502 554
82030-536PK 143.65 USD
82030-536 66016-086 66008-229 66016-158
Headspace Crimp Top Vials, SUN SRI™
Vials
Headspace vials manufactured of Type 1, Class B borosilicate glass.
Pre-assembled closures reduce contamination during sample preparation and save time in the laboratory.