PromoFluor, Maleimide, PromoKine
Supplier: PromoKine
PK-PF840-3-01 PK-PF594-3-01 PK-PF780-3-05 PK-PF505-3-01 PF510LSS-3-05 PF520LSS-3-01 PK-PF750-3-05 PF520LSS-3-05 PK-PF647P-3-05 PK-PF647P-3-01 PK-PF555P-3-01 PK-PF546-3-01 PK-PF488P-3-01 PK-PF568-3-01 PF510LSS-3-01 PK-PF546-3-05 PK-PF700P-3-01 PK-PF415-3-05 PF370LSS-3-01 PK-PF568-3-05 PK-PF488P-3-05 PK-PF770-3-05 PF488LSS-3-01 PK-PF670-3-05 PK-PF680-3-05 PK-PF633P-3-01 PF370LSS-3-05 PF375LSS-3-01 PF488LSS-3-05 PF380LSS-3-01 PF514LSS-3-05 PK-PF405-3-01 PK-PF594-3-05 PF500LSS-3-01 PF500LSS-3-05 PK-PF532-3-01 PF395LSS-3-01 PK-PF405-3-05 PK-PF555P-3-05 PF395LSS-3-05 PK-PF532-3-05 PK-PF680-3-01 PF350LSS-3-05 PK-PF770-3-01 PF350LSS-3-01 PK-PF840-3-05 PK-PF750-3-01 PK-PF780-3-01 PF514LSS-3-01 PK-PF415-3-01 PK-PF505-3-05 PK-PF670-3-01 PF375LSS-3-05 PK-PF633P-3-05 PF380LSS-3-05 PK-PF700P-3-05
10183-126EA 1635.43 USD
10183-126 10183-124 10183-044 10182-560 10182-286 10182-682 10182-880 10182-648 10182-604 10182-606 10183-042 10182-562 10182-684 10183-164 10183-162 10182-840 10182-842 10182-800 10182-494 10182-856 10182-614 10182-978 10183-090 10182-538 10182-616 10182-858 10182-816 10182-650 10182-496 10182-896 10182-776 10182-732 10182-898 10182-778 10182-734 10183-092 10183-106 10183-104 10183-146 10183-066 10182-582 10183-144 10182-706 10182-708 10182-980 10182-584 10183-064 10182-540 10182-878 10182-516 10182-956 10182-518 10182-754 10182-798 10182-954 10182-756
PromoFluor, Maleimide, PromoKine
Protein Linking and Labeling Systems
PromoFluor Maleimides are intended for covalently labeling biomolecules containing a thiol group (e.g. proteins).
  • Advantage over amino group labels
  • They do not interfere with techniques like differential 2D gel electrophoresis
  • Selectively modify a protein or peptide
  • Well-suited for labelling oligonucleotides
PromoFluor Maleimides are intended for covalently labeling biomolecules containing a thiol group (e.g
Maleimides couple to thiol groups under very mild conditions at a pH around 7. PromoFluor Dyes show a strong light absorption as well as high fluorescence intensity and photostability, good pH insensitivity and water solubility. The superior quality of our PromoFluor dyes and a very reasonable price making them an ideal alternative to many commonly used fluorophores.
proteins). A neutral thiol-reactive label has little influence on the isoelectric point of a protein. In effect, PromoFluor Maleimides have an advantage over amino group labels because they do not interfere with techniques like differential 2D gel electrophoresis. Since most proteins contain only a few thiol functional groups, thiol-reactive dyes can be used to selectively modify a protein or peptide containing a thiol group at a specific site. Moreover, these dyes are well-suited for labelling oligonucleotides.