Would you like to log in again?
PromoFluor, Amino-Modified Label, PromoKine
Supplier: Promocell, Inc.
PK-PF750-2-01 PF350LSS-2-01 PF520LSS-2-01 PK-PF670-2-01 PK-PF770-2-01 PF488LSS-2-01 PK-PF633P-2-01 PK-PF505-2-01 PK-PF488P-2-01 PF500LSS-2-01 PF514LSS-2-01 PF395LSS-2-01 PK-PF405-2-01 PF380LSS-2-01 PF375LSS-2-01 PK-PF532-2-01 PK-PF555P-2-01 PK-PF594-2-01 PK-PF700P-2-01 PK-PF546-2-01 PF510LSS-2-01 PK-PF568-2-01 PF370LSS-2-01 PK-PF780-2-01 PK-PF415-2-01 PK-PF680-2-01 PK-PF350-2-01 PK-PF647P-2-01
PromoFluor Amino-Modified Labels are intended for the covalent labeling of biomolecules containing a carboxy group e.g., glycoproteins.
  • Can be cross-coupled to carboxy groups through a mild in situ activation
  • Strong light absorption
  • Good pH insensitivity
  • Ideal alternative to many commonly used fluorophores
PromoFluor Amino-Modified Labels are intended for the covalent labeling of biomolecules containing a carboxy group e.g., glycoproteins. The amino group of PromoFluor Amino-Modified Labels can be cross-coupled to carboxy groups through a mild in situ activation. PromoFluor Dyes show a strong light absorption as well as high fluorescence intensity and photostability, good pH insensitivity and water solubility. The superior quality of our PromoFluor dyes and a very reasonable price making them an ideal alternative to many commonly used fluorophores.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
PromoFluor-350, amino-modified label 1 mg PK-PF350-2-01 10182-470 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-350LSS, amino-modified label 1 mg PF350LSS-2-01 10182-492 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-370LSS, amino-modified label 1 mg PF370LSS-2-01 10182-514 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-375LSS, amino-modified label 1 mg PF375LSS-2-01 10182-536 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-380LSS, amino-modified label 1 mg PF380LSS-2-01 10182-558 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-395LSS, amino-modified label 1 mg PF395LSS-2-01 10182-580 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-405, amino-modified label 1 mg PK-PF405-2-01 10182-602 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-415, amino-modified label 1 mg PK-PF415-2-01 10182-612 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-488 Premium, amino-modified label 1 mg PK-PF488P-2-01 10182-668 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-488LSS, amino-modified label 1 mg PF488LSS-2-01 10182-646 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500LSS, amino-modified label 1 mg PF500LSS-2-01 10182-730 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-505, amino-modified label 1 mg PK-PF505-2-01 10182-752 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-510LSS, amino-modified label 1 mg PF510LSS-2-01 10182-774 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-514LSS, amino-modified label 1 mg PF514LSS-2-01 10182-796 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-520LSS, amino-modified label 1 mg PF520LSS-2-01 10182-284 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-532, amino-modified label 1 mg PK-PF532-2-01 10182-838 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-546, amino-modified label 1 mg PK-PF546-2-01 10182-680 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-555P, Amino function 1 mg PK-PF555P-2-01 10182-854 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-568, Amino function 1 mg PK-PF568-2-01 10182-876 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-594, Amino function 1 mg PK-PF594-2-01 10182-894 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-633P, Amino function 1 mg PK-PF633P-2-01 10182-952 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-647P, Amino function 1 mg PK-PF647P-2-01 10182-976 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-670, amino-modified label 1 mg PK-PF670-2-01 10183-040 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-680, amino-modified label 1 mg PK-PF680-2-01 10183-062 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-700P, Amino function 1 mg PK-PF700P-2-01 10183-088 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-750, amino-modified label 1 mg PK-PF750-2-01 10183-102 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-770, amino-modified label 1 mg PK-PF770-2-01 10183-122 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-780, amino-modified label 1 mg PK-PF780-2-01 10183-142 Each (1mg) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty