Would you like to log in again?
Cytochrome c Apoptosis Detection Kit, PromoKine
Supplier: Promocell, Inc.
PK-CA577-K257 CA577-K1999-1
The PromoKine Cytochrome c Apoptosis Detection Kit provides an effective means for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis.
  • Provides an easy-to-use procedure
  • Unique reagents
  • Simple and straightforward procedure
Cytochrome c has been shown to play an important role in apoptosis. The protein is located in the space between the inner and outer mitochondrial membranes. An apoptotic stimulus triggers the release of cytochrome c from the mitochondria into the cytosol where it binds to Apaf-1. The cytochrome c/Apaf-1 complex activates caspase-9 which then activates caspase-3 and other downstream caspases of the apoptotic cascade. The PromoKine Cytochrome c Apoptosis Detection Kit provides an effective means for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis. The kit provides an easy-to-use procedure and unique reagents for separating a highly enriched mitochondria fraction from cytosol. Cytochrome c released from mitochondria into cytosol is then determined by Western blotting using the cytochrome c antibody provided in the kit. The procedure is very simple and straightforward - no ultracentrifugation is required and no toxic chemicals are involved.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Cytochrome c Apoptosis Detection Kit 100 assays CA577-K1999-1 10180-402 Each Retrieving Restricted
Magnetic Separator (re-usable) 1 PK-CA577-K257 10183-686 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty