Mitochondria Isolation Kit, PromoKine
Supplier: PromoKine
PK-CA577-K288
10183-710EA 609.28 USD
10183-710
Mitochondria Isolation Kit, PromoKine
Protein Extraction Protein Extraction Kits
PromoKine's Mitochondria Isolation Kit (for tissue & cultured cells) offers two options for the isolation of intact mitochondria.
  • Two options for the isolation of intact mitochondria
  • Utilize a reagent-based method
  • Traditional dounce homogenization
PromoKine's Mitochondria Isolation Kit (for tissue & cultured cells) offers two options for the isolation of intact mitochondria. The first option is utilizing a reagent-based method allowing parallel processing of multiple samples. The second option uses traditional dounce homogenization, which provides better mitochondrial yield.