Mammalian Whole Cell Protein Extraction Kit, PromoKine
Supplier: PromoKine
PK-CA577-K269
10183-704EA 451.9 USD
10183-704
Mammalian Whole Cell Protein Extraction Kit, PromoKine
Protein Extraction Protein Extraction Kits
The Mammalian Whole Cell Protein Extraction Kit has been optimized for complete extraction of all cellular proteins.
  • Optimized lysis buffer
  • Protease inhibitor cocktail
  • Entire procedure takes less than 20 minutes
The Mammalian Whole Cell Protein Extraction Kit has been optimized for complete extraction of all cellular proteins. It contains an optimized lysis buffer and a protease inhibitor cocktail as well as other reagents for convenient extraction of mammalian proteins from tissues and cells under non-denaturing conditions. The entire procedure takes less than 20 minutes, and the proteins can be used for many downstream applications such as gel electrophoresis, Western blotting, etc.