HDAC-11 Blocking Peptide
Supplier: BioVision
3611BP-50
10004-388EA 110.91 USD
10004-388
HDAC-11 Blocking Peptide
The peptide is used for blocking the antibody activity of HDAC-11 (Cat.# 3611P-100). It usually blocks the antibody activity completely in Western blot analysis by incubating the peptide with equal volume of antibody for 30 minutes at 37°C.
The peptide solution should be gently mixed before use.