HDAC-9 Blocking Peptide
Supplier: BioVision
3609BP-50
10004-382EA 110.91 USD
10004-382
HDAC-9 Blocking Peptide
The peptide is used for blocking the antibody activity of HDAC-9 (Cat.# 3609-100). It usually blocks the antibody activity completely in Western blot analysis by incubating the peptide with equal volume of antibody for 30 minutes at 37°C.
The peptide solution should be gently mixed before use.