Would you like to log in again?
PromoFluor Nick Translation Labeling Kit, PromoKine
Supplier: Promocell, Inc.
PF500-NTLK-10 PF660-PCRLK-10 PF500-DUTP-50 PF500-NTLK-50 PF-LK-546-05 PF430-PCRLK-50 PF430-DUTP-10 PF500-DUTP-10 PF430-DUTP-50 PF550-DUTP-10 PF640-DUTP-50 PF550-DUTP-50 PF640-DUTP-10 PF430-NTLK-50 PF430-NTLK-10 PF680N-DUTP-10 PF660-DUTP-50 PF500-PCRLK-10 PF680N-DUTP-50 PF500-PCRLK-50 PF660-DUTP-10 PF640-NTLK-10 PF-LK-546-10 PF430-PCRLK-10 PF640-NTLK-50 PF660-PCRLK-50 PF550-NTLK-50 PF550-NTLK-10
PromoKine’s PCR Labeling Kits and Nick Translation Labeling Kits have been designed for convenient direct DNA labeling by PCR or nick translation using Taq polymerase or Polymerase I/DNase I.
  • Optimized for enzymatic incorporation into DNA
  • Proprietary linker technology+
  • Ideal choice for all typical DNA labeling applications
PromoKine’s PCR Labeling Kits and Nick Translation Labeling Kits have been designed for convenient direct DNA labeling by PCR or nick translation using Taq polymerase or Polymerase I/DNase I
The Labeling Kits contain all reagents required for highly efficient, rapid, and easy-to-perform DNA labeling (excluding primers and template DNA).
The labeled dUTPs are also available separately.
The fluorescently labeled dUTP analogs provided with the kits have been optimized for enzymatic incorporation into DNA by using a proprietary linker technology. The high incorporation rates of the dye-dUTP analogs combined with the outstanding stability and quantum yield of the fluorophores make the kits the ideal choice for all typical DNA labeling applications such as FISH, single molecule detection, microarray gene expression profiling, and other nucleic acid hybridization assays.
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
PromoFluor-430 Nick Translation Labeling Kit 50 reactions PF430-NTLK-50 10182-630 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-430 Nick Translation Labeling Kit 10 reactions PF430-NTLK-10 10182-628 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-430 PCR Labeling Kit 50 reactions PF430-PCRLK-50 10182-634 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-430 PCR Labeling Kit 10 reactions PF430-PCRLK-10 10182-632 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-430-dUTP 50 µl PF430-DUTP-50 10182-626 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-430-dUTP 10 µl PF430-DUTP-10 10182-624 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500 Nick Translation Labeling Kit 50 reactions PF500-NTLK-50 10182-744 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500 Nick Translation Labeling Kit 10 reactions PF500-NTLK-10 10182-742 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500 PCR Labeling Kit 50 reactions PF500-PCRLK-50 10182-748 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500 PCR Labeling Kit 10 reactions PF500-PCRLK-10 10182-746 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500-dUTP 10 µl PF500-DUTP-50 10182-718 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-500-dUTP 50 µl PF500-DUTP-10 10182-716 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-546 Labeling Kit 5 reactions PF-LK-546-05 10183-186 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-546 Labeling Kit 10 reactions PF-LK-546-10 10183-188 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-550-dUTP 10 µl PF550-DUTP-10 10182-692 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-550-dUTP 50 µl PF550-DUTP-50 10182-694 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-550 Nick Translation Labeling Kit 10 reactions PF550-NTLK-10 10182-696 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-550 Nick Translation Labeling Kit 50 reactions PF550-NTLK-50 10182-698 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-640-dUTP 10 µl PF640-DUTP-10 10182-964 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-640-dUTP 50 µl PF640-DUTP-50 10182-966 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-640 Nick Translation Labeling Kit 10 reactions PF640-NTLK-10 10182-968 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-640 Nick Translation Labeling Kit 50 reactions PF640-NTLK-50 10182-970 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-660-dUTP 10 µl PF660-DUTP-10 10183-024 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-660-dUTP 50 µl PF660-DUTP-50 10183-026 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-660 PCR Labeling Kit 10 reactions PF660-PCRLK-10 10183-028 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-660 PCR Labeling Kit 50 reactions PF660-PCRLK-50 10183-030 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-680N-dUTP 10 µl PF680N-DUTP-10 10183-074 Each (10µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PromoFluor-680N-dUTP 50 µl PF680N-DUTP-50 10183-076 Each (50µl) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty