HDAC-5 Blocking Peptide
Supplier: BioVision
3605BP-50
10004-366EA 110.91 USD
10004-366
HDAC-5 Blocking Peptide
The peptide is used for blocking the antibody activity of HDAC-5 (Cat.# 3605-100). It usually blocks the antibody activity completely in Western blot analysis by incubating the peptide with equal volume of antibody for 30-60 minutes at 37°C.
The peptide solution should be gently mixed before use.