PromoKine's Cell Fractionation and Separation Kits contain unique formulations of buffers and reagents for convenient preparation of highly enriched cellular fractions from mammalian cells.
- Designed for serial sample preparation
- Convenient preparation of highly enriched cellular fractions
- Nuclear fractions from one sample
PromoKine's Cell Fractionation and Separation Kits contain unique formulations of buffers and reagents for convenient preparation of highly enriched cellular fractions (mitochondria, cytosol, nuclear) from mammalian cells. The Complete Cell Fractionation Kit is designed for serial sample preparation of four distinct protein fractions including cytosol, particulate, cytoskeleton, and nuclear fractions from one sample.