Fluorometric & Colorimetric ATP Quantitation Kit, PromoKine
Supplier: PromoKine
PK-CA577-K354
10180-426EA 777.43 USD
10180-426
Fluorometric & Colorimetric ATP Quantitation Kit, PromoKine
Colorimeters Comparator Kits
PromoKine's Fluorometric & Colorimetric ATP Quantitation Kit is designed to be a robust, simple method which utilizes the phosphorylation of glycerol to generate a product that is easily quantified by colorimetric or fluorometric methods.
  • Robust, simple method
  • Utilizes the phosphorylation of glycerol
  • Can detect as low as 50 picomol (1 µM) of ATP in various samples
ATP that is formed exclusively in the mitochondria is the major energy currency of living systems and virtually all energy requiring processes utilize the chemical energy stored in the phosphate bond of ATP. A variety of genetic diseases can affect ATP formation in the mitochondria. There are a number of commercially available ATP assays which detects femtomoles or less of ATP by measuring luminescence but these kits require specialized luminescence instrumentation and utilize luciferase which can be difficult to maintain in active form. PromoKine's Fluorometric & Colorimetric ATP Quantitation Kit is designed to be a robust, simple method which utilizes the phosphorylation of glycerol to generate a product that is easily quantified by colorimetric (λmax = 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The assay can detect as low as 50 picomol (1 µM) of ATP in various samples.