Fluorometric Cell Viability Kit I, PromoKine
Supplier: PromoKine
CA707-30025-1 CA707-30025-2 CA707-30025-0
10180-602EA 271.21 USD
10180-602 10180-604 10180-600
Fluorometric Cell Viability Kit I, PromoKine
Colorimeters Comparator Kits
Cell Viability Kit I provides a simple and rapid measurement of cell viability.
  • Single-step, homogenous cell proliferation assay
  • Sensitive, safe, and cost-effective
  • Requires as few as 80 cells for reproducible results
  • More sensitive, colorimetric measurement is also possible
The resazurin-based PromoKine Cell Viability Kit I provides a simple and rapid measurement of cell viability. Moreover, this single-step, homogenous cell proliferation assay is sensitive, safe, and cost-effective. It only requires as few as 80 cells for reproducible results. The redox dye Resazurin (also known as Alamar Blue) detects cell viability by converting from a non-fluorescent, blue dye to the highly red fluorescent dye resorufin in response to chemical reduction of the growth medium caused by cell growth. The fluorescent or colorimetric signal generated from the assay is proportional to the number of living cells in the sample. The kit reagents are nontoxic to cells and allow kinetic monitoring. While fluorometric resorufin quantitation is more sensitive, colorimetric measurement is also possible.