Conductive Vinyl Chair Mat, ACL Staticide
Supplier: ACL Staticide
6800
10856-362EA 214.08 USD
10856-362
Conductive Vinyl Chair Mat, ACL Staticide
Floor Mats Permanent Floor Mats
The ESD vinyl chair mat is made from engineered PVC with inherent electrical properties that will last the lifetime of the mat
Made in the USA. 100% recyclable.
The mat features a stippled surface, with a UV additive to protect color and eliminate fading. Resistance is 10³ to 10 ohms.
Certifications: Compliant to ANSI/ESDA S20.20. Passes weathering ozone resistance per ASTE D2240.