Would you like to log in again?
Stericheck Dilutor, MDI
Supplier: MDI MEMBRANE TECHNOLOGIES, INC.
SVPD0902XXX209 SVPD0902XXX309
The Stericheck Dilutor is a presterilized accessory for dissolution of difficult to dissolve powders or dilution of viscous fluids. The dilutor is specifically designed to work with difficult to dissolve powders or viscous fluid in a vial which are required to be diluted for testing.
  • Non-coring needle design
  • Pre-installed color coded clamps
  • Raised transparent vent
  • Customized needle connections
  • Validated for equal sample distribution
  • Compatible with Steripump or other available sterility testing equipment
The Stericheck Dilutor features a single needle with expansion chamber and vent, and a double needle with PVC tubing
ETO Sterilized models can be used under a laminar flow hood. Gamma Sterilized models can be used in isolators or laminar flow hoods.
Raised vents prevent air flow obstruction due to fluid logging, and 'L' shaped flow directors prevent frothing. A tough membrane helps withstand inadvertent back pressure.
Certifications: Validated as per USP 71 Sterility Tests. Validation Guide available on request.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Stericheck Dilutor, ETO Sterilized SVPD0902XXX209 75792-102 Case of 40 Retrieving
Pack of 20 Retrieving
Stericheck Dilutor, Gamma Sterilized SVPD0902XXX309 75792-104 Case of 40 Retrieving
Pack of 20 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty